Learn More
Enzo Life Sciences HSP60 (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADIEKS600
Encompass_Preferred
The HSP60 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis. Alternative name: Heat shock protein 60. Assay Time: <3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-200-7216
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.