Learn More
Enzo Life Sciences AMPIGENE HS Taq Mix (1000Reactions)
Supplier: Enzo Life Sciences ENZNUC1011000
AMPIGENE HS Taq Mix uses the latest developments in polymerase technology and buffer chemistry to optimize and increase PCR sensitivity. The AMPIGENE HS Taq Mix is a robust mix using hot-start technology to increase sensitivity for PCR applications including genotyping, multiplex PCR, screening, library construction, colony PCR and PCR direct from blood and urine. An optimized buffer system allows efficient amplification under fast and standard cycling conditions. The AMPIGENE HS Taq Mix is resistant to PCR inhibitors and suitable for direct PCR from unprocessed samples: bacterial culture, bacterial colonies, blood, and urine. Applications: PCR.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.