Learn More
Hach Company HQ411D Laboratory Single Input, pH/mV Meter - pH and ORP
Supplier:  Hach Company HQ411D
Encompass_Preferred
Designed for water quality experts, the Hach HQ411d Laboratory pH Meter is an advanced laboratory digital meter that takes the guesswork out of measurements. HQD laboratory meters connect with smart Intellical electrodes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced HQD digital meter is designed to withstand years of continuous lab use. HQD bench meters feature a simple user interface that does not require manuals or training to operate. The HQ411d pH-meter is ideal for measuring pH, or mV/RedOx/ORP, in wastewater, drinking water or general water quality, soil, and food or beverages applications. A wide range of Intellical electrodes and sensors are available to perform accurate pH measurements in any sample media types.
Catalog No. 50-199-7711
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.