Learn More
Enzo Life Sciences PATHO-GENE HPV type 16 18 31 33 51 probe (1ml)
Supplier: Enzo Life Sciences ENZ328821000
The PATHO-GENE HPV type 16/18/31/33/51 probe is a mixture of biotin-labeled HPV-16, HPV- 18, HPV-31, HPV-33 and HPV-51 specific probes in buffered formamide and hybridization enhancers. Long Term Storage: +4°C.