Learn More
Foxx Life Sciences EZWaste® HPLC VersaCap®GL45 Solvent Waste System, Cap-Only W/ Exhaust Filter, Six (6x) Ports for OD Tubing 3.2mm (1/8"") or 1.6mm (1/16""), 1/EA
SDP
Supplier:  Foxx Life Sciences 3300D11FLS
Encompass_Preferred
Foxx Life Sciences EZWaste® HPLC VersaCap® GL45 Solvent Waste Cap Assembly is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from multiple HPLC machines. This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers and provides safer mechanism of disposal for vapor wastes. The EZwaste® Solvent Waste system also features a closed loop design which protects against leaks and spills, improving overall lab safety.Each EZwaste® Assembly includes a VersaCap® GL45 with Manifold, fittings 6 x 3.2 mm (1/8") OD Tubing, fittings 6 x 1.6 mm (1/16") OD Tubing, 6 x Plugs suitable for blocking of unused ports & 1 carbon exhaust filter. * Depending on application, carbon exhaust filter life cycle may vary.
Catalog No. 50-172-0598
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.