Learn More
Foxx Life Sciences EZWaste® HPLC Universal Stackable 38-430 Solvent Waste System, Cap-Only W/ Exhaust Filter, Six(6x) Ports OD Tubing -3.2 mm (1/8"") or 1.6 mm (1/16""), Three (3x) Hose Barb 3.2mm (1/8"") or 9.5mm(3/8""), Three (3x) OD Tubing 12.7 mm (1/2""), 1/EA
Supplier: Foxx Life Sciences 3300103FLS