Learn More
Foxx Life Sciences PUREGRIP® Universal HPLC Solvent Reservoir VersaCap® GL45 System with Class VI PTFE Adapter & Safety Air Inlet Valve, Cap-Only, Six (6x) Ports- OD Tubing-3.2 mm (1/8"") or 1.6 mm (1/16""), 1/EA
Supplier: Foxx Life Sciences 4500600FLS