Learn More
Air Science Usa Horizontal Clean Bench 60" Wide with UV System
Supplier: Air Science Usa HLF60A(UV)
Horizontal Laminar Flow Cabinet 60" Wide With UV Lamp and front cover. Protects your non-hazardous applications with horizontal unidirectional (laminar) ISO Class 4 air across the work area. Includes Stainless Steel Workurface. CART-BW-HLF-60 Base Stand avilable and sold separatly.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.