Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBS Scientific Horizontal Midi-Gel Productivity Packages
Allows casting second gel while the first is running for increased efficiency.
Specifications
Type | Midi-Gel System |
---|---|
Certifications/Compliance | CE |
Warranty | 3 Years |
Description | Compatible Gel Tray: WGT-500 |
Catalog Number | Mfr. No. | Gel Dimensions | Description | Volume (Metric) Buffer | Dimensions (L x W x H) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Gel Dimensions | Description | Volume (Metric) Buffer | Dimensions (L x W x H) | Price | Quantity & Availability | ||
S22884
|
CBS Scientific
GCMHU502 |
L x W: 10 x 14 cm (3.9 x 5.5 in.) | Compatible Gel Tray: WGT-500 | 700 mL | 21.5 x 16.7 x 14 cm (8.4 x 6.5 x 5.5 in.) | N/A | N/A | N/A | |
S22885
|
CBS Scientific
GCMHU602 |
L x W: 16 x 14 cm (6.2 x 5.5 in.) | Compatible Gel Tray: WGT-600 | 750 mL | 27.5 x 16.7 x 14 cm (10.8 x 6.5 x 5.5 in.) | N/A | N/A | N/A | |
Description
Midi Horizontal Productivity Packages feature platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling.- Platinum electrode cassettes can be easily removed for cleaning or replacement
- High quality, heavy duty construction with vast array of accessory options
- Winged gel trays for easy handling. No more sticking fingers into slippery buffer to extract gel tray
- Addition of external gel caster and additional gel tray allows researcher to use time more efficiently
- MHU-602 Midi-Horizontal runs up to a full microtitre plate (108 samples) per gel
- Tapered baffles allow easy in situ gel formation with or without gel tray
- Run up to 32 samples per gel. Two slots in each gel tray
- Combs are printed with catalog number and thickness for easy identification
- Choice of double-sided adjustable height combs gives researcher more options
- Optional PTFE-coated aluminum combs for low-friction comb removal and accelerated well formation
- Horizontal Midi-Gels are available in four vivid colors; green, yellow, blue or fluorescent pink
- Color-coded horizontal systems are recommended to differentiate exclusive use by specific lab or for specific purpose
Specifications
Midi-Gel System | |
CE | |
3 Years | |
Compatible Gel Tray: WGT-500 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title