Learn More
Honeywell™ PRO 5007 Structural Firefighting Bunker Boots - C Width
Superior combination of comfort and absorbency
$1109.89 - $1114.82
Specifications
Height (English) 14 in.
View More Specs
Products 15
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-821-407
View Documents
Honeywell™
BT50070901
9C
Pair for $1,114.82
Add to Cart
18-821-403
View Documents
Honeywell™
BT50070701
7C
Pair for $1,114.82
Add to Cart
18-821-405
View Documents
Honeywell™
BT50070801
8C
Pair for $1,114.82
Add to Cart
18-821-406
View Documents
Honeywell™
BT50070851
8.5C
Pair for $1,114.82
Add to Cart
18-821-402
View Documents
Honeywell™
BT50070651
6.5C
Pair for $1,114.82
Add to Cart
18-821-401
View Documents
Honeywell™
BT50070601
6C
Pair for $1,109.89
Add to Cart
18-821-404
View Documents
Honeywell™
BT50070751
7.5C
Pair for $1,114.82
Add to Cart
18-821-408
View Documents
Honeywell™
BT50070951
9.5C
Pair for $1,114.82
Add to Cart
18-821-409
View Documents
Honeywell™
BT50071001
10C
Pair for $1,114.82
Add to Cart
18-821-410
View Documents
Honeywell™
BT50071051
10.5C
Pair for $1,114.82
Add to Cart
18-821-411
View Documents
Honeywell™
BT50071101
11C
Pair for $1,114.82
Add to Cart
18-821-413
View Documents
Honeywell™
BT50071201
12C
Pair for $1,114.82
Add to Cart
18-821-412
View Documents
Honeywell™
BT50071151
11.5C
Pair for $1,114.82
Add to Cart
18-821-414
View Documents
Honeywell™
BT50071301
13C
Pair for $1,114.82
Add to Cart
18-821-415
View Documents
Honeywell™
BT50071401
14C
Pair for $1,114.82
Add to Cart
Description
Description
  • Quilted to 7.5 oz. Kevlar™ felt thermal barrier and laminated to fully encapsulating Crosstech™ Footwear Fabric bootie
  • Crosstech™ moisture barrier provides superior liquid penetration resistance to blood, body fluids and NFPA tested common chemicals
  • Military AB leather construction insulates and adds abrasion-, puncture-, water- and chemical-resistance
  • With heavy duty 0.75 in. reinforced straps mounted to the exterior of boot and anchored with a 2 × 1.5 in. shield-shaped base
  • External texon board shinguard protects and guards sensitive shin area of legs
  • Patented PowerToe™ cover resists toe abrasion and provides unparalled wear over the lifetime of the boots
  • Oblique toe safety cap provides additional room in height and width of boot
  • Fire/Ice PowerSole™ angled lugs provide substantial edging and gripping capabilities
  • Molded thermoplastic heel counter locks foot into the boot and anchors it in place
  • Patented PowerHeel™ cover protects heel area from abusive doffing of boots
  • Corrison-resistant steel ladder shank and stainless steel, full length, toe-to-heel puncture resistant bottom plate
Compliance
Meet NFPA 1971, 2007 edition requirements
Specifications
Specifications
14 in.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.