Learn More
Promocell GmbH PromoCell hMoCD14+-PB-c 10 million Human CD14+ Monocytes, derived from Peripheral Blood, single donor, cryopreserved
Supplier:  Promocell GmbH C12909
Encompass_Preferred
Monocytes (Mo) are immature phagocytic cells circulating in the blood. Monocytes can differentiate into several different cell types. The most common applications for Monocytes are the in vitro differentiation into dendritic cells (moDC) and osteoclasts as well as their use as phagocytic immune cells.CD14 belongs to the family of LPS receptor antigens and is strongly expressed on the majority of Monocytes. Thus, binding of immunomagnetic particles to this antigen is employed for purification of PromoCell CD14+-Monocytes (hMoCD14+-PB) from Mononuclear Cells (MNC).PromoCell Human CD14+ Monocytes are isolated from fresh peripheral blood. After isolation of the ultra-pure mononuclear cell fraction by proprietary methods, the CD14+ Monocytes are isolated by positive selection with 95% purity.Our hMoCD14+ -PB are now also available from HLA-typed donors.
Catalog No. 50-306-073
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.