Learn More
Innovative Research Inc Hmn tPA 85 Sngl Chn Pur 1mg
Supplier: Innovative Research Inc IHUTPA85SC1MG
Human tPA 85% Single Chain Purified 1mg - Human tPA 85% Single Chain Purified from Innovative Research has been recombinantly produced in CHO cells and purified using affinity chromatography. This is a frozen liquid buffered in 0.4M HEPES; 0.1M NaCl; pH 7.4 and a purity of >95%. This product is useful as a positive control for ELISA and western blot and for a variety of in vitro applications including cell differentiation, cell signaling, clotting assays, fibrinolysis, plasminogen activation, protein-protein interactions, receptor binding, thromboelastography, and thrombolysis. Fully active, >85% single chain. Synthesized from cDNA from a human melanoma cell line. More Details Source Chinese Hamster Ovary Cell Culture Purification Affinity purified Purity >95% Storage Conditions -70C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.