Learn More
Innovative Research Inc Hmn Prorenin Ntv Rcmb 100ug
Supplier:  Innovative Research Inc IHUPRENNR100UG
Encompass_Preferred
Human Prorenin Native Recombinant 100ug - Human Prorenin Native Recombinantis recombinantly produced in HEK cells and purified by affinity chromatography. This is a Frozen liquid buffered in 0.05M Tris-HCl; 0.05M NaCl; pH 8.0 and a purity of 95% as determined by SDS-PAGE. More Details Species Human Target Prorenin Purification Method Affinity purified Purity 95% as determined by SDS-PAGE Analysis Source Human Embryonic Kidney cells Storage Conditions -70C; Protect from light Additional Information Untagged native form prorenin is recombinantly produced in HEK cell culture and purified using affinity chromatography. Can be fully activated to renin using catalytic amounts of trypsin. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including cell signaling, hypertension studies, protein-protein interactions, receptor binding, and more.
Catalog No. 50-274-4264
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.