Learn More
Electron Microscopy Sciences Holey Carbon Film with Ultrathin Carbon < 3 nm thick on 400 mesh Nickel Grid
Supplier:  Electron Microscopy Sciences HC400NICC
Encompass_Preferred
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Carbon < 3 nm. Supported by a mesh grid.
Grid: Nickel
Catalog No. 50-193-1377
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.