Learn More
Electron Microscopy Sciences Holey Carbon Film with Ultrathin Carbon < 3 nm thick on 400 mesh Gold Grid
Supplier: Electron Microscopy Sciences HC400AUCC
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Carbon < 3 nm. Supported by a mesh grid.
Grid: Gold