Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DR Mouse anti-Human, BUV395, Clone: L243, BD Horizon™

Mouse Monoclonal Antibody

$166.00 - $436.00

Specifications

Antigen HLA-DR
Clone L243
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Ultraviolet 395
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB568647
View Documents
BD Biosciences
568647
100 Tests
Each for $436.00
Add to Cart
 
BDB568648
View Documents
BD Biosciences
568648
25 Tests
Each for $166.00
Add to Cart
 
Description

Description

HLA-DR Monoclonal antibody specifically detects HLA-DR in Human samples. It is validated for Flow cytometry
Specifications

Specifications

HLA-DR
Flow Cytometry
Brilliant Ultraviolet 395
Mouse
Human
MHC class II antigen; HLA class II histocompatibility antigen
IgG2a κ
Affinity Chromatography
L243
Monoclonal
Liquid
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
Human lymphoblastoid B-cell line RPMI 8866
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.