Learn More
Electron Microscopy Sciences Histo-Mark Pen
Supplier:  Electron Microscopy Sciences 7215512
Encompass_Preferred
The Histo-Mark pen is designed to work like a pen. It is a fine-line marker that can be used on any surface. The Histo-Mark pen is solvent resistent, especially to alcohol, acetone, xylene, xylene subsite and toluene. Marking quality remains consistent throughout the life of the ink barrel.
Catalog No. 50-366-032
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.