Learn More
New England Biolabs, Inc. HiScribe™ T7 ARCA mRNA Kit – 20 reactions
SDP
Supplier:  New England Biolabs, Inc. E2065S
Encompass
Most eukaryotic mRNAs require a 7-methyl guanosine (m7G) cap structure at the 5' end and a Poly(A) tail at the 3 end to be efficiently translated. By using a DNA template encoding a poly(A) tail, the HiScribe T7 ARCA mRNA Kit can be used to synthesize capped and tailed mRNAs. The cap structure is added to the mRNA by co-transcriptional incorporation of Anti-Reverse Cap Analog (ARCA) using T7 RNA Polymerase. Poly(A) tail is incorporated during the transcription reaction. The kit also includes DNase I and LiCl for DNA template removal and quick mRNA purification. Additionally, the kit is capable of partial incorporation of modified UTP and CTP (up to 50% each) without affecting the mRNA yield significantly. By using a DNA template encoding a poly(A) tail, capped and tailed modified mRNA can be synthesized in a single reaction in 30 minutes. mRNAs synthesized with the kit can be used for cell transfection, microinjection, in vitro translation and RNA vaccines.
Catalog No. 50-104-7498
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.