Learn More
Enzo Life Sciences HIGHDEF yellow IHC chromogen (HRP) (30 ml)
SDP
Supplier:  Enzo Life Sciences ADI9501700030
Encompass_Preferred
Unique high definition yellow IHC chromogen for HRP-activated development. HIGHDEF yellow IHC chromogen (HRP) is intended to be used in conjunction with horseradish peroxidase (HRP)-based immunostaining or in situ hybridization systems. With at least 1 day working stability, this substrate/chromogen system can be used in any automation system as well as manual use. Contents: HIGHDEF yellow IHC chromogen (HRP) substrate buffer and chromogen, Empty mixing bottle. Application: IHC, ISH (in situ hybridization). Long Term Storage: +4°C
Catalog No. NC0504414
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.