Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBS Scientific High-Throughput Submarine Horizontal Electrophoresis Systems
SCI_ED

Run up to 816 samples with these units.

Supplier:  CBS Scientific HSU2614

 View more versions of this product

Catalog No. S22906


Description

Description

CBS Economy Horizontal Productivity Packages feature platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling.

Gel Tray Specifications:
  • Winged gel trays for easy handling - no more sticking fingers into slippery buffer to extract gel tray
  • Made from sturdy 6.4mm thick UV transparent acrylic
  • Grooves in gel tray corners create agarose anchors to prevent gel flotation during electrophoresis
  • Gel tray configuration allows single run or multiple simultaneous short runs
  • Three options for gel casting; tapered baffles, external gel casters or continuously adjustable gel caster
  • All combs are adjustable height and have an aluminum backing to keep the combs perfectly straight during gel casting with hot agarose
  • All combs come with microtitre format to allow multi-channel pipetting for accuracy and speed
  • Comb profiles are diagrammed at right to show many configurations, with or without markers
  • PTFE-coated combs allow faster well formation and are recommended when using low percentage or low melt agarose
  • Buffer recirculation ports allow maintenance of uniform pH and ionic strength and reduce frequency of buffer changes necessary during long runs.
  • Tight-fitting safety cover reduces condensation during electrophoresis
  • Coolant can be recirculated beneath gel bed to provide constant temperature runs
  • Combs are printed with catalog number and thickness for easy identification
Applications: Rapid Screening fo cloning products, rapid screening of restriction fragments, rapid screening of synthetic oligos, rapid screening of PCR products
Specifications

Specifications

CE
Compatible Gel Tray: WGT-2614
L x W: 14 x 26 cm (5.5 x 10.2 in.)
3 Years
38 x 28 x 10 cm (14.9 x 11 x 3.9 in.)
Blue
Electrophoresis System
2200 mL
Gel Tray I.D.: 14 x 24.7 cm (5.5 x 9.7 in.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.