Learn More
Worldwide Glass Resource High Recovery Vials
Vials engineered for automated systems
Supplier: Worldwide Glass Resource MER0010331
Description
- Manufactured from highly-resistant Type I borosilicate glass
- Clear in color
Specifications
Clear | |
Borosilicate Glass | |
2 in. | |
3.1 mL | |
51 mm |
15 mm | |
3.1 mL | |
80 | |
Screw Cap | |
80 Cs. |