Learn More
World Precision Instrument High Intensity Fiber Optic Illumination Source-Z-LITE-Z186
SDP
Supplier:  World Precision Instrument ZLITEZ186
Encompass_Preferred
The Z-LITE Fiber Optic Il­lu­mi­na­tor provides reli­able, un­in­ter­rupt­ed high-intensity light for micro­scopes. Z-LITE allows a continuous range of subdued or concentrated light­ing controlled by a rotary dim­mer on the front panel. Z-LITE may be used with a ring light and single or bi­fur­cat­ed flex­i­ble fiber bundles, en­abling the light beam to be placed ex­act­ly where it is need­ed. Forced air cool­ing prolongs the lamp life. Lamp color temper­a­ture is 3350°K. An interlock switch automatically cuts off power when front panel is opened to replace bulb.
Catalog No. 50-253-8250
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.