Learn More
FUJIFILM IRVINE SCIENTIFIC INC HEPES Buffer Solution 1M, 100ml
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 9319100ML
Encompass
HEPES Buffer Solution 1M is designed for use as a buffering agent for cell culture. This formulation contains  238.3 HEPES g/L in water, pH 7.2 to 7.4 at room temperature.
Catalog No. NC0786742
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.