Learn More
Andwin Scientific HEMACYTOMETER COVER SLIDES
Supplier: Andwin Scientific 48300470
The hemacytometer cover glass is manufactured from a clean, noncorrosive glass. This product measures 20 x 26 millimeters and complies with the federal regulations. It is sold in a pack of 12.