Learn More
Andwin Scientific HEMACYTOMETER COVER GLASS 0.4
SDP
Supplier:  Andwin Scientific 6351430
Encompass_Preferred
The Hemacytometer cover glass measures 20 x 26 mm. It has a thickness of 0.5 mm.
Catalog No. NC9443751
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.