Learn More
Eurofins Discoverx PathHunter HEK 293 PTGIR β-Arrestin Cell Line, 2 vials. (93-0305C1)
Supplier: Eurofins Discoverx 930305C1
The PathHunter PTGIR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin I2, beraprost) based activation of PTGIR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTGIR and EA-tagged β-Arrestin-2. Activation of PTGIR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.