Learn More
Eurofins Discoverx PathHunter HEK 293 PTGER4 β-Arrestin Cell Line, 2 vials. (93-0304C1)
Supplier:  Eurofins Discoverx 930304C1
Encompass_Preferred
The PathHunter PTGER4 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin E2) based activation of PTGER4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTGER4 and EA-tagged β-Arrestin-2. Activation of PTGER4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0214
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.