Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Radleys Carousel Reaction Station Accessory, Reaction Station Insert
SDP

Accessory and replacement part for Radleys Carousel 6 Plus™ and Carousel 12 Plus™ Reaction Stations

Supplier:  Heidolph™ 015861250

Catalog No. 05-403-851


Description

Description

For use with Radleys Carousel 6 Plus (Cat. No. 13-889-503) and Radleys Carousel 12 Plus (Cat. No. 13-889-501) Reaction Stations

Specifications

Specifications

Reaction Station Insert
Radleys Carousel 6 Plus™ and Carousel 12 Plus™ reaction system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.