Learn More
Heidolph™ Radleys Jacketed Vessel Kit
SDP
Radleys jacketed vessel kit for use with Reactor-Ready™ and Reactor-Ready™ Duo lab reactors
Supplier:  Heidolph™ 015950111
 View more versions of this product
Catalog No. 05-405-263
Description
Description
  • These cost effective vessel kits include the vessel and accessories you need to get started.
  • The Pt100 probe and anchor stirrer included in each kit are matched to the vessel size.
  • Each vessel kit includes reaction vessel, PTFE anchor stirrer, side arm couplings, Pt100 PTFE temperature probe with LEMO connector, and PTFE temperature probe adapter.
Specifications
Specifications
446°F
5000 mL
Reactor-Ready™ and Reactor-Ready™ Duo Lab Reactors
Single jacketed
Jacketed Vessel Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.