Learn More
Chemglass Life Sciences 500mL Glas-Col Heating Mantle, Series M, Spherical, 270 Watts
Supplier: Chemglass Life Sciences CG1100007
500mL Glas-Col Heating Mantle, Series M, Spherical, 270 Watts. The Glas-Col Series M mantle is the most compact mantle with an aluminum shell that can heat flasks up to 72 liters. Like the series TM, this mantle has a durable aluminum housing outside and resilient glass fabric inside to support flasks and protect flask walls from thermal strain. The internal operating temperature is 450 C. Only the bottom of the flask is covered so the contents being heated are fully visible. The mantle is grounded through the aluminum housing and requires a support such as CG-11004 through CG-11008. All mantles are 115 Volt and are supplied with a detachable 4 foot, 3 wire cord.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.