Learn More
Chemglass Life Sciences HEATING MANTLE 1000ML
SDP
Supplier:  Chemglass Life Sciences CG1250007
Encompass_Preferred
Mantle, Heating; Chemglass; Glas-Col; Series STM; Used in distillation, fractionation processes; w/Splash guard and poncho safety shield; For high temp. operations; Upper internal operating temp.: 650 deg. C; 1000mL; 115V, 600 watts
Catalog No. 50-401-783
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.