Learn More
Cole-Parmer BriskHeat TBIH051-080LD Dual Element, Cloth Insulated Heat Tape, 8' x 1/2" , 624 Watts, 120 V
Supplier: Cole-Parmer 3605030
Dual-element design improves the heater’s flexibility over standard high-temperature tapes. Tapes are made of two heating elements that are knitted together in series and then wound together with high-temperature Samox fiberglass. The 120 VAC tapes include a plug Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.