Specifications
Empty Storm Case | |
HPX High Performance Resin, Multilayer Cubed Foam | |
31.3 x 20.4 x 12.2 in. (79.5 x 51.82 x 30.99 cm) | |
10.5 in. | |
26.67 cm | |
31.3 in. | |
79.5 cm | |
29 lb. | |
20.4 in. | |
51.82 cm |
Orange | |
5 press and pull latches, 2 padlockable hasps, flush powerful hinges | |
12.2 in. | |
30.99 cm | |
Three soft-grip handles, in-line wheels, telescoping handle, vortex valve | |
29 in. | |
73.66 cm | |
13.15 kg | |
18 in. | |
45.72 cm |