Specifications
Cubed Storm Case | |
HPX High Performance Resin, Multilayer Cubed Foam | |
24.6 x 19.7 x 8.6 in. (62.48 x 50.04 x 21.84 cm) | |
8 in. | |
20.3 cm | |
24.6 in. | |
62.48 cm | |
16 lb. | |
19.7 in. | |
50.04 cm |
Orange | |
4 press and pull latches, 2 padlockable hasps, flush hinges | |
8.6 in. | |
21.84 cm | |
Double-layered, soft-grip handle, vortex valve | |
22 in. | |
55.8 cm | |
7.26 kg | |
17 in. | |
43.1 cm |