Learn More
AVANTOR MASTERFLEX (VWR LLC) Hand-operated siphon pump, 5 GPM, Polyethylene
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0642220
Encompass_Preferred
Siphon pumps are inexpensive, disposable hand pumps. Model 06439-00 features a squeeze-bulb action while models 06422-20 and 06439-10 feature a bellows action for quick and easy emptying of containers. Model 07079-70 creates a siphon with a few strokes of the pump.
Catalog No. NC1208569
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.