Learn More
Proteintech Group Inc Halo-Trap Magnetic Agarose Kit
Supplier: Proteintech Group Inc OTMAK20
The ChromoTek Halo-Trap Magnetic Agarose Kit consists of an anti-Halo-tag Nanobody (VHH), which is covalently bound to magnetic agarose beads. Halo-Trap Magnetic Agarose Kit is used to immunoprecipitate Halo-tag proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc. in the absence or presence of a covalently bound ligand. The interaction between Halo-Trap and the Halo-tag protein is reversible.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.