Learn More
Proteintech Group Inc Halo-Trap Agarose beads
Supplier: Proteintech Group Inc OTA200
Halo-Trap Agarose are affinity beads for IP of Halo-tag proteins. It comprises a Halo-tag Nanobody /VHH conjugated to agarose beads.
This item is not returnable.
View return policy