Learn More
Enzo Life Sciences H1-7 (1mg). CAS: 65189-70-0
Supplier: Enzo Life Sciences BMLP1060001
This sequence, RRKASGP, is derived from calf thymus histone H1 containing Ser38. Peptide is phosphorylated by protein kinase A. Alternative name: Histone H1 phosphorylation site, PKA substrate, Arg-Arg-Lys-Ala-Ser-Gly-Pro, RRKASGP. Purity: ≥97%. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.