Learn More
Thermo Scientific Dionex™ CarboPac™ PA200 IC Columns for Dual EGC Mode
Supplier:  Thermo Scientific 302862
Encompass
Type Guard Column | Length (Metric) 50 mm | Particle Size 5.5 µm | Diameter (Metric) 1.0 mm | Max. Pressure 4500 psi (at 0.063 mL/min.) | Temperature (Metric) 4°C to 60°C | For Use With (Application) Complex Carbohydrate Analysis in Dual EGC Mode | Column Construction PEEK Resin Composition 5.5 µm diameter ethylvinylbenzene/divinylbenzene substrate (55% cross-linked) agglomerated with 43 nm microbead quaternary amine functionalized latex (6% cross-linked). | For Use With (Equipment) Dionex CarboPac PA200, 1.0 x 250 mm Analytical Column
Catalog No. NC1706237
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.