Learn More
Bioworld GRIFFONIA SIMPLICIFOLIA- LE
Supplier:  Bioworld 215100051
Encompass_Preferred
GRIFFONIA SIMPLICIFOLIA- LECTIN: Girffonia simplicifolia Lectin isolated from Griffonia simplicifolia. Specifications: Primary sugar specificity: Galactose/N-acetylgalactosamine Preferential binding activity: A-rich: alpha-N-acety...
Catalog No. 50-198-736
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.