Learn More
Andwin Scientific GREEN LASER CRYO-BABIES LAR
SDP
Supplier:  Andwin Scientific 91871702
Encompass_Preferred
The 1.28" L x 0.5" W Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196�C to 100�C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in the color of green.
Catalog No. 50-821-014
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.