Learn More
CBS Scientific Gradient Markers
SCI_ED
Linear with side outlets.
Supplier:  CBS Scientific GM1000
 View more versions of this product
Catalog No. S05788
Description
Description
  • Gradient flow is controlled by a PTFE-coated valve centered between the two chambers
  • The outflow channel is fitted with a male luer adapter which can be used with any needle or luer valve
Specifications
Specifications
Electrophoresis
Gradient Marker
16.9 oz.
500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.