Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ GPT HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Human overexpression lysate
Supplier: Abnova™ L118T6
Specifications
HEK293 | |
GPT HEK293 Cell Transient Overexpression Lysate(Non-Denatured) | |
Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. | |
pCMV-GPT full length | |
100 μg | |
Human | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunoprecipitation, Western Blot | |
BC018207 | |
Theoretical M.W. 55kDa | |
Transient overexpression cell lysate was tested with anti-GPT antibody (Mfr. No. H00002875-M03) by Western Blot. | |
In modified RIPA Lysis Buffer. | |
2 mg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction