Learn More
Crescent Chemical Co Inc GLUTARALDEHYDE 25%
SDP
Supplier:  Crescent Chemical Co Inc SE23114.01
Encompass_Preferred
Glutaraldehyde 25 % solution in water for electron microscopy, high purity In vials; filled under argon. Size - 25ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 23114.01
Catalog No. 50-247-655
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.