Learn More
World Precision Instrument Gluck Bone Shears, Semi Box Eccentric Lock, 8.5 in
SDP
Supplier:  World Precision Instrument WP0019
Encompass_Preferred
Gluck Rib Shears make cutting through ribs and small bones quick and easy, especially in thoracic and cardiovascular applications. The curved blade lets you properly position the shears, and the locking mechanism gives you control of the grip at the tip. The beveled end minimizes surrounding tissue trauma, and the tip design allow for access in confined spaces. Made of standard grade stainless steels, these bone cutters may be disinfected or autoclaved between uses.
Catalog No. 50-253-8382
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.