Learn More
Eisco™ Glass Burette, Sure Trace Certification
Class A, DIN 385 Compliant, Borosilicate Glass with Glass Key Stopcock.
Supplier:  Eisco™ CH0236DST
 View more versions of this product
Catalog No. 12-183-074
Explore all special offers
Add to Cart
Description
Description
  • Accurate as per class A, DIN ISO 385 Compliant
  • Includes an attached key stopcock made of high quality glass
  • Burette is made of precisely bore high quality, heavy duty Borosilicate 3.3 glass. Built to last
  • Features chemical resistant blue printing. All contents are included in a single package
Specifications
Specifications
100 mL
Borosilicate Glass (3.3)
Class A
82.55 cm
Buret
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.