Learn More
Proteintech Group Inc GFP-Trap Magnetic Particles M-270 Kit
Supplier: Proteintech Group Inc GTDK20
GFP-Trap Magnetic Particles M-270 for immunoprecipitation (IP) of GFP-tagged proteins. GFP-Trap Magnetic Particles M-270 is highly recommended, when very large proteins/complexes are investigated, and magnetic separation is needed for IP. It consists of a GFP VHH/ Nanobody coupled to Magnetic Particles M-270.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.