Learn More
Proteintech Group Inc GFP-Trap Magnetic Agarose
SDP
Supplier:  Proteintech Group Inc GTMA20
Encompass
The ChromoTek GFP-Trap Magnetic Agarose are affinity beads for IP of GFP-fusion proteins. It comprises a GFP Nanobody/ VHH coupled to magnetic agarose beads.
Catalog No. 50-225-8342
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.