Learn More
Proteintech Group Inc GFP-Booster ATTO488
Supplier: Proteintech Group Inc GBA48810
anti-GFP VHH/ Nanobody conjugated to a fluorescent dye for IF/ microscopy.
This item is not returnable.
View return policy