Learn More
ATCC Mumps virus RNA; Strain: Enders
Supplier:  ATCC VR106D
Encompass
Genomic RNA isolated from a preparation of cell lysate and supernatant from SL-29 cells (ATCC CRL-1590™) infected with Mumps virus, strain Enders (ATCC VR-106™). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. Source organism and host cells are also available through the ATCC Catalog.
Catalog No. NC2290376
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.